missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Magnesium, Reference Standard Solution (1000ppm ±1%/Certified), Fisher Chemical

$84.88 - $119.88

Chemical Identifiers

CAS 13446-18-9
Molecular Formula H12MgN2O12
Molecular Weight (g/mol) 256.403
MDL Number MFCD00085308
InChI Key MFUVDXOKPBAHMC-UHFFFAOYSA-N
PubChem CID 202877
IUPAC Name magnesium;dinitrate;hexahydrate
SMILES [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.[Mg+2]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
SM51-500 SDS View Documents Thermo Scientific
SM51500
500mL Poly Bottle Each for $119.88
Case of 6 Each for $719.28
SM51-100 SDS View Documents Thermo Scientific
FLSM51100
100mL Poly Bottle Each for $84.88
Case of 6 Each for $509.28
Specifications

Chemical Identifiers

13446-18-9
256.403
MFUVDXOKPBAHMC-UHFFFAOYSA-N
magnesium;dinitrate;hexahydrate
H12MgN2O12
MFCD00085308
202877
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.[Mg+2]

Specifications

13446-18-9,7697-37-2,7732-18-5
990 to 1010ppm
1000ppm ±1%
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.[Mg+2]
256.403
Liquid
Poly Bottle
14mmHg at 20°C
100°C
500mL
H12MgN2O12
MFCD00085308
MFUVDXOKPBAHMC-UHFFFAOYSA-N
magnesium;dinitrate;hexahydrate
202877
Certified
LOT NUMBER USED Magnesium ref. Std.
0°C
Colorless
Magnesium, Reference Standard Solution, 1000ppm ±1%
Safety and Handling

Safety and Handling

DOTInformation : DOT Class 8, : Corrosive