Thermo Scientific™ M-20 Microplate Swinging Bucket Rotor
Spin up to six microplates and, using sealed carriers, ensure certified sealed conditions for all samples with the Thermo Scientific™ M-20 Microplate Swinging-Bucket Rotor.
Manufacturer:  Thermo Scientific™ 75003624
Catalog No. 75-003-624
Description
Description
Spin up to six microplates and, using sealed carriers, ensure certified sealed conditions for all samples with the Thermo Scientific™ M-20 Microplate Swinging-Bucket Rotor.
Ordering Information:
Order 75003625 for Sealed Carriers, with trays and lids (set of 2), and 75002011 for Replacement Sealing Lids (set of 2). For Replacement O-rings for lids (set of 4 with grease) order 75002012. Contact your Sales Representative for information.
Includes:
Two buckets, two adapters
Compatible with:
Thermo Scientific Sorvall™ 16/16R, Sorvall Lengend™ X1/X1R, Heraeus™ Multifuge™ X1, Heraeus Megafuge™ 16 Centrifuge Series
Specifications
Specifications
M-20 Microplate Rotor
2 x 3 Standard or 2 x 1 Deepwell Plates
4,000rpm
Thermo Scientific™ Sorvall™ Legend™ XT/XF, Sorvall Legend™ X1, Sorvall ST 40, Sorvall ST 16, Heraeus™ Multifuge™ X3/X3F, Heraeus™ Multifuge™ X1, Heraeus™ Megafuge™ 40, Heraeus™ Megafuge™ 16 Centrifuge Series.
Biocontainment certified by CAMR™, HPA, Porton Down, U.K.
Swinging Bucket Rotor
2,272 x g
12.7cm
90°
Documents