Learn More
Lithium salicylate, tech., Thermo Scientific Chemicals
$63.11 - $506.26
Chemical Identifiers
CAS | 552-38-5 |
---|---|
Molecular Formula | C7H5LiO3 |
Molecular Weight (g/mol) | 144.054 |
MDL Number | MFCD00045813 |
InChI Key | PSBOOKLOXQFNPZ-UHFFFAOYSA-M |
Synonym | lithium salicylate, unii-93f1sp6qin, salicylic acid lithium salt, dilithium salicylate, 93f1sp6qin, lithium 2-hydroxybenzoate, lithium 1+ ion salicylate, lithiumsalicylate, lithiumsalicylat, salicylicacidlithiumsalt |
PubChem CID | 23663621 |
IUPAC Name | lithium;2-hydroxybenzoate |
SMILES | [Li+].C1=CC=C(C(=C1)C(=O)[O-])O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AA3935614
|
Thermo Scientific Chemicals
03935614 |
25 g |
Each for $63.11
|
|
|||||
AA3935622
|
Thermo Scientific Chemicals
03935622 |
100 g |
Each for $167.22
|
|
|||||
AA3935636
|
Thermo Scientific Chemicals
03935636 |
500 g |
Each for $506.26
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
552-38-5 | |
144.054 | |
PSBOOKLOXQFNPZ-UHFFFAOYSA-M | |
23663621 | |
[Li+].C1=CC=C(C(=C1)C(=O)[O-])O |
C7H5LiO3 | |
MFCD00045813 | |
lithium salicylate, unii-93f1sp6qin, salicylic acid lithium salt, dilithium salicylate, 93f1sp6qin, lithium 2-hydroxybenzoate, lithium 1+ ion salicylate, lithiumsalicylate, lithiumsalicylat, salicylicacidlithiumsalt | |
lithium;2-hydroxybenzoate |
Specifications
552-38-5 | |
Sweet | |
MFCD00045813 | |
lithium salicylate, unii-93f1sp6qin, salicylic acid lithium salt, dilithium salicylate, 93f1sp6qin, lithium 2-hydroxybenzoate, lithium 1+ ion salicylate, lithiumsalicylate, lithiumsalicylat, salicylicacidlithiumsalt | |
PSBOOKLOXQFNPZ-UHFFFAOYSA-M | |
lithium;2-hydroxybenzoate | |
23663621 | |
Technical | |
Lithium salicylate |
(decomposition) | |
C7H5LiO3 | |
25 g | |
Soluble in water | |
[Li+].C1=CC=C(C(=C1)C(=O)[O-])O | |
144.054 | |
144.05 | |
Powder |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H302-H314-H335
EINECSNumber : 209-011-7
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only