Learn More
Lithium bis(trimethylsilyl)amide, 0.9-1.1M in hexane, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals
$129.44 - $478.85
Chemical Identifiers
CAS | 4039-32-1 |
---|---|
Molecular Formula | C6H18LiNSi2 |
Molecular Weight (g/mol) | 167.327 |
MDL Number | MFCD00008261 |
InChI Key | YNESATAKKCNGOF-UHFFFAOYSA-N |
Synonym | lithium bis trimethylsilyl amide, lithium hexamethyldisilazide, lihmds, lhmds, hexamethyldisilazane lithium salt, unii-rc4n1i108m, lithiumbis trimethylsilyl amide, lithium bis trimethylsilyl azanide, lithium hexamethyldisilazane, lithium bis-trimethylsilyl amide |
PubChem CID | 2733832 |
IUPAC Name | lithium;bis(trimethylsilyl)azanide |
SMILES | [Li+].C[Si](C)(C)[N-][Si](C)(C)C |
Description
Lithium bis(trimetylsilyl)amide is used as a strong base in organic synthesis and as catalysts in polymerization reactions. It is also used to form various organolithium compounds such as acetylides and lithium enolates. It is a base employed in generating enolates for preparation of lactone precursors, pyranones and cyclohexanes. Further, it is used to catalyze the addition of phosphine P-H bonds to carbodiimides leading to phosphaguanidines. In addition, it is used in a three-step synthesis of disubstituted 1,2,5-thiadiazoles.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Packaged under argon
Chemical Identifiers
4039-32-1 | |
167.327 | |
YNESATAKKCNGOF-UHFFFAOYSA-N | |
2733832 | |
[Li+].C[Si](C)(C)[N-][Si](C)(C)C |
C6H18LiNSi2 | |
MFCD00008261 | |
lithium bis trimethylsilyl amide, lithium hexamethyldisilazide, lihmds, lhmds, hexamethyldisilazane lithium salt, unii-rc4n1i108m, lithiumbis trimethylsilyl amide, lithium bis trimethylsilyl azanide, lithium hexamethyldisilazane, lithium bis-trimethylsilyl amide | |
lithium;bis(trimethylsilyl)azanide |
Specifications
4039-32-1 | |
0.716 g/mL | |
ChemSeal™ bottles | |
[(CH3)3Si]2NLi | |
100 g | |
8136861 | |
lithium bis trimethylsilyl amide, lithium hexamethyldisilazide, lihmds, lhmds, hexamethyldisilazane lithium salt, unii-rc4n1i108m, lithiumbis trimethylsilyl amide, lithium bis trimethylsilyl azanide, lithium hexamethyldisilazane, lithium bis-trimethylsilyl amide | |
YNESATAKKCNGOF-UHFFFAOYSA-N | |
lithium;bis(trimethylsilyl)azanide | |
2733832 | |
Liquid |
Colorless to Yellow | |
-31°C (-24°F) | |
C6H18LiNSi2 | |
MFCD00008261 | |
UN2924 | |
Air and moisture sensitive | |
Soluble in tetrahydrofuran,hexane and toluene. | |
[Li+].C[Si](C)(C)[N-][Si](C)(C)C | |
167.327 | |
167.33 | |
Lithium bis(trimethylsilyl)amide |
Safety and Handling
GHS H Statement
H225-H304-H361-H373-H314-H336
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Suspected of damaging fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Causes severe skin burns and eye damage.
May cause drowsiness or dizziness.
P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
H225-H304-H314-H335-H336-H361f-H373
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
TSCA : Yes
storageNote1 :
- Packaged under argon
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only