missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Lithium 2-ethylhexanoate, Thermo Scientific Chemicals
$46.30 - $443.15
Chemical Identifiers
CAS | 15590-62-2 |
---|---|
Molecular Formula | C8H15LiO2 |
Molecular Weight (g/mol) | 150.146 |
MDL Number | MFCD00058702 |
InChI Key | MAZKNBXUMANNDL-UHFFFAOYSA-M |
Synonym | lithium 2-ethylhexanoate, 2-ethylhexanoic acid lithium salt, hexanoic acid, 2-ethyl-, lithium salt 1:1, hexanoic acid, 2-ethyl-, lithium salt, acmc-20akn3, lithium 1+ ion 2-ethylhexanoate, lithium 1+ 2-ethylhexanoate, hexanoic acid,2-ethyl-, lithium salt 1:1 |
PubChem CID | 23672311 |
IUPAC Name | lithium;2-ethylhexanoate |
SMILES | [Li+].CCCCC(CC)C(=O)[O-] |
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
15590-62-2 | |
150.146 | |
MAZKNBXUMANNDL-UHFFFAOYSA-M | |
23672311 | |
[Li+].CCCCC(CC)C(=O)[O-] |
C8H15LiO2 | |
MFCD00058702 | |
lithium 2-ethylhexanoate, 2-ethylhexanoic acid lithium salt, hexanoic acid, 2-ethyl-, lithium salt 1:1, hexanoic acid, 2-ethyl-, lithium salt, acmc-20akn3, lithium 1+ ion 2-ethylhexanoate, lithium 1+ 2-ethylhexanoate, hexanoic acid,2-ethyl-, lithium salt 1:1 | |
lithium;2-ethylhexanoate |
Specifications
15590-62-2 | |
MFCD00058702 | |
lithium 2-ethylhexanoate, 2-ethylhexanoic acid lithium salt, hexanoic acid, 2-ethyl-, lithium salt 1:1, hexanoic acid, 2-ethyl-, lithium salt, acmc-20akn3, lithium 1+ ion 2-ethylhexanoate, lithium 1+ 2-ethylhexanoate, hexanoic acid,2-ethyl-, lithium salt 1:1 | |
[Li+].CCCCC(CC)C(=O)[O-] | |
150.146 | |
150.15 | |
Lithium 2-ethylhexanoate |
C8H15LiO2 | |
2 g | |
MAZKNBXUMANNDL-UHFFFAOYSA-M | |
lithium;2-ethylhexanoate | |
23672311 | |
Lumps |
Safety and Handling
GHS H Statement
H361
Suspected of damaging fertility or the unborn child.
P201-P202-P281-P308+P313-P501c
H361d
EINECSNumber : 239-657-5
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only