Learn More
Lithium 2,4-pentanedionate, 99.5%, Thermo Scientific Chemicals
$39.85 - $335.82
Chemical Identifiers
CAS | 18115-70-3 |
---|---|
Molecular Formula | C5H7LiO2 |
Molecular Weight (g/mol) | 106.05 |
MDL Number | MFCD00013496 |
InChI Key | JTEOOCAGEXVCBQ-LNKPDPKZSA-M |
Synonym | Lithium acetylacetonate |
IUPAC Name | lithium(1+) (2Z)-4-oxopent-2-en-2-olate |
SMILES | [Li+].C\C([O-])=C\C(C)=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AA3052909
|
Thermo Scientific Chemicals
03052909 |
10 g |
Each for $39.85
|
|
|||||
AA3052918
|
Thermo Scientific Chemicals
03052918 |
50 g |
Each for $100.66
|
|
|||||
AA3052930
|
Thermo Scientific Chemicals
03052930 |
250 g |
Each for $335.82
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
18115-70-3 | |
106.05 | |
JTEOOCAGEXVCBQ-LNKPDPKZSA-M | |
lithium(1+) (2Z)-4-oxopent-2-en-2-olate |
C5H7LiO2 | |
MFCD00013496 | |
Lithium acetylacetonate | |
[Li+].C\C([O-])=C\C(C)=O |
Specifications
18115-70-3 | |
White | |
MFCD00013496 | |
4575087 | |
Lithium acetylacetonate | |
[Li+].C\C([O-])=C\C(C)=O | |
106.05 | |
99.5% | |
Lithium 2,4-pentanedionate |
∼250°C (decomposition) | |
C5H7LiO2 | |
10 g | |
Hygroscopic | |
JTEOOCAGEXVCBQ-LNKPDPKZSA-M | |
lithium(1+) (2Z)-4-oxopent-2-en-2-olate | |
106.05 | |
Crystalline |
Safety and Handling
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P201-P202-P261-P264b-P270-P271-P280i-P281-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335-H361
EINECSNumber : 242-008-9
TSCA : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only