Invitrogen™ Countess™ II FL Automated Cell Counter DFS Item
Cell counting and viability in < 10 seconds with no need to buy slides or tips. Two optional user-changeable fluorescence channels with over 20 light cubes to choose from
Manufacturer:  Invitrogen™ LSAMQAF1000
Catalog No. AMQAF1000
Includes: Countess USB drive for storing and transferring data to your computer, Disposable slide holder, Reusable slide holder, Light cube removal tool, Power cord and 4 plug adaptors to accommodate most countries, Quick reference card (QRC), A free box of 50 disposable slides
Description
Description
The new Countess™ II FL Automated Cell Counter is a benchtop assay platform equipped with state-of-the-art optics, full autofocus, and image analysis software for rapid assessment of cells in suspension. With three-channel flexibility—brightfield and two optional fluorescence channels—researchers can count cells, monitor fluorescent protein expression, evaluate apoptosis, and measure cell viability.
  • Accurate—use the Countess II FL Automated Cell Counter to eliminate the subjectivity of manual cell counting and user-to-user variability
  • Flexible—the Countess II FL Automated Cell Counter is able to accept up to two EVOS™ light cubes that are user-selectable from over twenty fluorescence channels
  • Fast—the Countess II FL Automated Cell Counter provides a total cell count, reports viability, and measures average cell size in as little as 10 seconds. Counting speed is identical when using fluorescence channels.
  • Convenient—the Countess II FL Automated Cell Counter requires no cleaning or routine maintenance. A capacitive touchscreen and simple user interface provides quick startup and requires minimal training.
The Countess II FL Automated Cell Counter comes with:
  • Countess USB drive for storing and transferring data to your computer
  • Disposable slide holder
  • Reusable slide holder
  • Light cube removal tool
  • Power cord and 4 plug adaptors to accommodate most countries
  • Quick reference card (QRC)
  • A free box of 50 disposable slides
The Countess II FL Automated Cell Counter works in 3 simple steps:
  1. Mix 10 µL of sample with 10 µL of trypan blue and pipet into a disposable Countess chamber slide.
  2. Insert the slide into the instrument.
  3. Press the “Count” button.
Get accurate cell counts
The Countess II FL Automated Cell Counter uses trypan blue staining combined with an auto-focus mechanism and sophisticated image analysis algorithm to obtain accurate cell and viability counts. The measurement range extends from 1 x 10e4 to 1 x 10e7 cells/mL (the optimal range is 1 x 10e5 to 4 x 10e6 cells/mL), which is broader than that of a hemocytometer. The optimal cell size for accurate viability assessment is 7–60 µm, while particles as small as 4 µm can be successfully counted.
Simple operation with fluorescent application versatility
Operated using an intuitive user interface, automated cell counting with the Countess II FL instrument gives you the data you need to prepare your sample for the next passage or experiment, and allows you to multiplex with a variety of fluorescent dyes and proteins excited via EVOS™ Light Cubes.
Transfer data and images to your PC
Using the USB port on the Countess II FL Automated Cell Counter, you can save your cell count results and images, and then transfer them to your PC. (For your convenience, a USB drive is supplied with the Countess II FL instrument).
Specifications
Specifications
One 2-chamber disposable or 2-chamber reusable slide
139.7 mm
9 in.
Countess USB drive for storing and transferring data to your computer, Disposable slide holder, Reusable slide holder, Light cube removal tool, Power cord and 4 plug adaptors to accommodate most countries, Quick reference card (QRC), A free box of 50 disposable slides
Cell Size: 4-60 µm (detection), 7-60 µm (viability)
8 lb.
228.6 mm
5.5 in.
7' Capacitive touch screen
228.6 mm
USB Stick
100-240 VAC
9 in.
Documents
Promotions
Promotions
For research use only.  Not for use in diagnostic procedures.