missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MP Biomedicals™ Leibovitz L-15 Medium

Used for growth of tissues in free gaseous exchange with the atmosphere.

Manufacturer:  MP Biomedicals™ 091251054

Catalog No. MP91251054



Description

Description

In this medium, dextrose is replaced by galactose. In addition, buffering is provided by the free bases of the amino acids in place of sodium bicarbonate. As a result this medium has been used for growth of tissues in free gaseous exchange with the atmosphere.

Specifications

Specifications

Sterile
Mammalian Cell Culture
500mL
Store at 0° to 5°C
Leibovitz L-15 Medium
Liquid
Filtration
L-glutamine
Documents