Learn More
Lead, plasma standard solution, Specpure™ Pb 1000μg/mL, Thermo Scientific Chemicals
$88.22 - $289.75
Chemical Identifiers
CAS | 7439-92-1 |
---|---|
Molecular Formula | Pb |
Molecular Weight (g/mol) | 207.20 |
MDL Number | MFCD00134050 |
InChI Key | WABPQHHGFIMREM-UHFFFAOYSA-N |
PubChem CID | 5352425 |
ChEBI | CHEBI:27889 |
IUPAC Name | lead |
SMILES | [Pb] |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AA13853AD
|
Thermo Scientific Chemicals
013853AD |
50 mL |
Each for $88.22
|
|
|||||
AA13853AE
|
Thermo Scientific Chemicals
013853AE |
100 mL |
Each for $125.75
|
|
|||||
AA13853AP
|
Thermo Scientific Chemicals
013853AP |
500 mL |
Each for $289.75
|
|
|||||
Description
Lead, plasma standard solution is used as a standard solution in analytical chemistry and atomic absorption spectroscopy. It is also used as a single-element standard solution for plasma emission spectrometry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7439-92-1 | |
207.20 | |
WABPQHHGFIMREM-UHFFFAOYSA-N | |
CHEBI:27889 | |
[Pb] |
Pb | |
MFCD00134050 | |
5352425 | |
lead |
Specifications
7439-92-1,7697-37-2,7732-18-5 | |
Colorless | |
MFCD00134050 | |
[Pb] | |
207.20 | |
CHEBI:27889 | |
23 hPa (17mm Hg) at 20°C | |
Liquid |
50 mL | |
Pb | |
WABPQHHGFIMREM-UHFFFAOYSA-N | |
lead | |
5352425 | |
Nitric acid: 5.0%; Lead: 0.1%; Water: 94.9% | |
Miscible with water | |
Lead |
Safety and Handling
P201-P202-P234-P260-P264b-P271-P281-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c
H290-H314-H335-H350-H360-H373
DOTInformation : Hazard Class: 8; Packaging Group: II
EINECSNumber : 231-714-2
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only