missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Labtainer™ BPC Bag with 3 Ports, MPC Insert, MPC Body, and Luer Lock

Using these BPC bags with 3 ports (MPC insert, MPC body, luer lock), simplify cleaning and disposal and reduce cross-contamination risks for 2 to 50L volumes of media, buffers, water for injection (WFI) and biological products.

$102.03 - $189.45

Specifications

Certifications/Compliance cGMP
Description Two-dimensional disposable cell culture/bioprocess bag
Film Description CX5-14 Single Web Film
Port Detail 3/8in. Quick Disconnect Insert; 3/8in. Quick Disconnect Body; 1/8in. Female Luer Lock
Sterilization Method Gamma Irradiated
View More Specs

Products 5
Catalog Number Mfr. No. Capacity (Metric) Quantity Price Quantity  
Catalog Number Mfr. No. Capacity (Metric) Quantity Price Quantity  
SH3071201 View Documents Thermo Scientific™
SH3071201
5L 1 Pack for $110.52
SH3071202 View Documents Thermo Scientific™
SH3071202
10L 1 Pack for $117.22
SH3071203 View Documents Thermo Scientific™
SH3071203
20L 1 Pack for $122.15
SH3071204 View Documents Thermo Scientific™
SH3071204
50L 1 Pack for $189.45
SH3071205 View Documents Thermo Scientific™
SH3071205
2L 1 Each for $102.03
Description

Description

Using Thermo Scientific™ Labtainer™ bags for 2 to 50L volumes, simplify cleaning and disposal and reduce cross-contamination risks and start up costs.

Labtainers are constructed with our CX5-14 single web, multi-layer cast film manufactured in a cGMP facility. Our standard Labtainers are suitable for a wide array of bioprocess applications. For optimal fit, form and function, Labtainers can be customized using the industry's largest component library.

Highlights:

  • 11 sizes from 50mL to 50L
  • Film designed to incorporate best combination of flexibility and durability for collection, storage and transport of sterile bioprocess liquids: superior stress resistance, puncture resistance and physical properties to ensure stability of contents
  • Gamma-irradiation sterilized
  • Convenient end-port design with a handle at the opposing end
  • Space-efficient, ergonomic alternative to rigid walled carboys
  • Customizable via the industry's largest library of qualified components

Recommended for:

  • Sampling – collection of in-process samples at a variety of bioprocessing stages

  • Harvest – collection and storage of bulk product from bioreactors

  • Storage – sterile storage of media, buffers and process intermediates

  • Separation – feeding and receiving of sterile liquids from separation system

  • Purification – buffer feeding to purification system, fraction collection and storage

  • Filling – sterile reservoir for filling systems

  • Transport Shipping – bulk sterile liquids, buffers, media and process intermediates within and between facilities

Specifications

Specifications

cGMP
CX5-14 Single Web Film
Gamma Irradiated
No
Two-dimensional disposable cell culture/bioprocess bag
3/8in. Quick Disconnect Insert; 3/8in. Quick Disconnect Body; 1/8in. Female Luer Lock
Ambient