missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Labconco™ Protector™ Standard Storage Cabinet
The Labconco Protector™ Standard Storage Cabinet may be used for storage of supplies and small laboratory instruments.
$2933.80 - $3813.93
Specifications
For Use With (Application) | Ideal for storage and venting of small laboratory instrument and mild acid and other reagent |
---|---|
Includes | Four Leveling Feet and One 8 in. D Filler Panel to Increase Cabinet Depth from 22 to 30 in. |
Load Bearing Capacity (English) | 800 lb. |
Product Type | Storage Cabinet |
Material | Epoxy-coated Steel |
Catalog Number | Mfr. No. | Width (English) | Door Type | Weight (English) | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Width (English) | Door Type | Weight (English) | Price | Quantity | |||
16-305-50
|
Labconco™
9900300 |
24 in. | Single, Right Hinge, Manual Close | 130 lb. |
Each for $3,235.32
|
|
|||
16-305-51
|
Labconco™
9900600 |
24 in. | Single, Left Hinge, Manual Close | 130 lb. |
Each for $3,235.32
|
|
|||
16-305-80
|
Labconco™
9900000 |
48 in. | Dual, Manual Close | 260 lb. |
Each for $3,813.93
|
|
|||
16-305-79
|
Labconco™
9900100 |
36 in. | Dual, Manual Close | 180 lb. |
Each for $3,479.80
|
|
|||
16-305-81
|
Labconco™
9900200 |
30 in. | Dual, Manual Close | 155 lb. |
Each for $3,307.20
|
|
|||
16-305-48
|
Labconco™
9900400 |
18 in. | Single, Right Hinge, Manual Close | 90 lb. |
Each for $3,096.78
|
|
|||
16-305-46
|
Labconco™
9900500 |
12 in. | Single, Right Hinge, Manual Close | 65 lb. |
Each for $2,933.80
|
|
|||
16-305-49
|
Labconco™
9900700 |
18 in. | Single, Left Hinge, Manual Close | 90 lb. |
Each for $3,096.78
|
|
|||
16-305-47
|
Labconco™
9900800 |
12 in. | Single, Left Hinge, Manual Close | 65 lb. |
Each for $2,933.80
|
|
|||
16-305-67
|
Labconco™
9904000 |
24 in. | Single, Right Hinge, Manual Close | 120 lb. |
Each for $3,430.92
|
|
|||
16-305-64
|
Labconco™
9904100 |
18 in. | Single, Right Hinge, Manual Close | 85 lb. |
Each for $3,324.97
|
|
|||
16-305-63
|
Labconco™
9904200 |
12 in. | Single, Right Hinge, Manual Close | 60 lb. |
Each for $3,145.68
|
|
|||
16-305-66
|
Labconco™
9904400 |
18 in. | Single, Left Hinge, Manual Close | 85 lb. |
Each for $3,324.97
|
|
|||
16-305-68
|
Labconco™
9904300 |
24 in. | Single, Left Hinge, Manual Close | 120 lb. |
Each for $3,430.92
|
|
|||
16-305-65
|
Labconco™
9904500 |
12 in. | Single, Left Hinge, Manual Close | 60 lb. |
Each for $3,145.68
|
|
|||
Description
The Labconco Protector™ Standard Storage Cabinet may be used for storage of supplies and small laboratory instruments.
- Support loads up to 800 lb. (363kg)
- Durable epoxy-coated steel construction
- Attractive glacier white exterior complements laboratory casework and Labconco fume hoods
- Manual-closing, non-locking door(s)
Order Info
Work surface and interior shelf kits are sold separately
Specifications
Ideal for storage and venting of small laboratory instrument and mild acid and other reagent | |
800 lb. | |
Epoxy-coated Steel |
Four Leveling Feet and One 8 in. D Filler Panel to Increase Cabinet Depth from 22 to 30 in. | |
Storage Cabinet |