missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Labconco™ Pan Cart

Holds a wide variety of general lab equipment

Supplier:  Labconco™ 8047500

Catalog No. 11-925-20


Description

Description

  • Load capacity: 400 lb. (181kg)
  • Strong, versatile lab cart made of 1 in. (2.5cm) powder-coated square steel tubing frame
  • Welded crossbars under the center of the pans provide support and minimize vibration noise
  • Two watertight, 2 in. deep (5cm) powder-coated steel pans with ergonomic, extruded rubber handle grips
  • Removable gray rubber mats line pans, prevent loads from shifting
  • 4 in. dia. (10cm) nonmarking casters; two with toe locks

Order Info

Weight: 50 lb. (23kg).

Warranty

Lifetime guarantee.

Specifications

Specifications

Glacier white
Epoxy-coated
48.3 cm
34.88 x 19.0 x 36.38 in. (88.6 x 48.3 x 92.4 cm)
Two 2 in. (51 mm) deep epoxy-coated steel, removable watertight pans, lined with removable gray rubber mats; 4 in. diameter casters, two with toe locks; Extruded rubber handle grips; Epoxy-coated 18 gauge welded tubular steel frame
SDS
Documents

Documents

Promotions

Promotions