missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
L-Tyrosine methyl ester, 98%, Thermo Scientific Chemicals
$100.19 - $276.16
Chemical Identifiers
CAS | 1080-06-4 |
---|---|
Molecular Formula | C10H13NO3 |
Molecular Weight (g/mol) | 195.22 |
MDL Number | MFCD00002392 |
InChI Key | MWZPENIJLUWBSY-VIFPVBQESA-N |
Synonym | l-tyrosine methyl ester, h-tyr-ome, tyrosine methyl ester, methyl l-tyrosinate, tyr-ome, methyl tyrosinate, tyrosine, methyl ester, s-2-amino-3-4-hydroxy-phenyl-propionic acid methyl ester, methyl 2s-2-amino-3-4-hydroxyphenyl propanoate, s-methyl 2-amino-3-4-hydroxyphenyl propanoate |
PubChem CID | 70652 |
ChEBI | CHEBI:17215 |
IUPAC Name | methyl (2S)-2-amino-3-(4-hydroxyphenyl)propanoate |
SMILES | COC(=O)[C@@H](N)CC1=CC=C(O)C=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH6206914
|
Thermo Scientific Chemicals
H6206914 |
25 g |
Each for $100.19
|
|
|||||
AAH6206922
|
Thermo Scientific Chemicals
H6206922 |
100 g |
Each for $276.16
|
|
|||||
Description
L-Tyrosine methyl ester is used as a dehydroepiandrosterone as a sulfate acceptor
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsL-Tyrosine methyl ester is used as a dehydroepiandrosterone as a sulfate acceptor
Solubility
Soluble in water.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Chemical Identifiers
1080-06-4 | |
195.22 | |
MWZPENIJLUWBSY-VIFPVBQESA-N | |
70652 | |
methyl (2S)-2-amino-3-(4-hydroxyphenyl)propanoate |
C10H13NO3 | |
MFCD00002392 | |
l-tyrosine methyl ester, h-tyr-ome, tyrosine methyl ester, methyl l-tyrosinate, tyr-ome, methyl tyrosinate, tyrosine, methyl ester, s-2-amino-3-4-hydroxy-phenyl-propionic acid methyl ester, methyl 2s-2-amino-3-4-hydroxyphenyl propanoate, s-methyl 2-amino-3-4-hydroxyphenyl propanoate | |
CHEBI:17215 | |
COC(=O)[C@@H](N)CC1=CC=C(O)C=C1 |
Specifications
134°C to 136°C | |
C10H13NO3 | |
2372626 | |
Soluble in water. | |
COC(=O)[C@@H](N)CC1=CC=C(O)C=C1 | |
195.22 | |
CHEBI:17215 | |
98% | |
L-Tyrosine methyl ester |
1080-06-4 | |
MFCD00002392 | |
l-tyrosine methyl ester, h-tyr-ome, tyrosine methyl ester, methyl l-tyrosinate, tyr-ome, methyl tyrosinate, tyrosine, methyl ester, s-2-amino-3-4-hydroxy-phenyl-propionic acid methyl ester, methyl 2s-2-amino-3-4-hydroxyphenyl propanoate, s-methyl 2-amino-3-4-hydroxyphenyl propanoate | |
MWZPENIJLUWBSY-VIFPVBQESA-N | |
methyl (2S)-2-amino-3-(4-hydroxyphenyl)propanoate | |
70652 | |
195.22 | |
25 g |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 214-095-3
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only