missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

L-Histidine (White Crystals or Crystalline Powder), Fisher BioReagents

Manufacturer:  Fisher BioReagents FLBP382100

Catalog No. BP382-100




Description

Description

This amino acid is suitable for use in tissue culture systems requiring additives.
Specifications

Specifications

L-Histidine
71-00-1
10ppm max.
98.5 to 101.0 %
C6H9N3O2
15, 4758
l-histidine, histidine, h-his-oh, glyoxaline-5-alanine, anti-rheuma, l---histidine, istidina, s-histidine, l-histidin, l-histidine
Not Detected Detected/Not Detect.
(2S)-2-amino-3-(1H-imidazol-5-yl)propanoic acid
6274
CHEBI:15971
Solid
98.5 to 101.0%
White
7.0 to 8.5
0.02% max.
1ppm max.
0.10% max.
Pass Test
0.20% max.
Glass Bottle
HNDVDQJCIGZPNO-YFKPBYRVSA-N
C1=C(NC=N1)CC(C(=O)O)N
155.157
12.0 to 13.0° (+ or -)
155.16
98.0% min.
Histidine
282°C
100g

Chemical Identifiers

71-00-1
155.157
l-histidine, histidine, h-his-oh, glyoxaline-5-alanine, anti-rheuma, l---histidine, istidina, s-histidine, l-histidin, l-histidine
CHEBI:15971
C1=C(NC=N1)CC(C(=O)O)N
C6H9N3O2
HNDVDQJCIGZPNO-YFKPBYRVSA-N
6274
(2S)-2-amino-3-(1H-imidazol-5-yl)propanoic acid
Safety and Handling

Safety and Handling

Recommended Storage : RT

SDS Documents
Promotions

Promotions