missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

L-Glutamine (White Crystals or Crystalline Powder), Fisher BioReagents

Manufacturer:  Fisher BioReagents BP379100

Catalog No. BP379-100



 

Description

Description

This amino acid is suitable for use in tissue culture systems requiring additives.
Specifications

Specifications

L-Glutamine
56-85-9
10ppm max.
Pass Test
98.5 to 101.0 %
0.2% max.
15, 4507
ZDXPYRJPNDTMRX-VKHMYHEASA-N
(2S)-2,5-diamino-5-oxopentanoic acid
146.146
CHEBI:18050
Solid
Glutamine
185°C
100g
0.1% max.
1ppm max.
0.1% max.
98.5 to 101.0%
C5H10N2O3
1.0% max.
Glass Bottle
C(CC(=O)N)C(C(=O)O)N
6.3 to 7.3° (+ or -)
5961
146.15
95.0% min.
White
4.0 to 6.0

Chemical Identifiers

56-85-9
146.146
5961
C5H10N2O3
ZDXPYRJPNDTMRX-VKHMYHEASA-N
CHEBI:18050
Safety and Handling

Safety and Handling

Recommended Storage : RT

SDS Documents