DWK Life Sciences Kimble™ Kontes™ Lids for Rectangular TLC Developing Tanks
Latches hold lid firmly to the chromatography tank. Fits 20 x 20mm chromatography developing tanks.
Manufacturer:  DWK Life Sciences 4161910000
Catalog No. K416191-0000
Description
Description
  • Latch lids have a knob for easy removal and PTFE retainers
  • Nonslip lids have a knob for easy lifting and PTFE retainers inside to prevent the lid from accidentally sliding off the tank
Specifications
Specifications
Fits 20 x 20mm chromatography developing tank
PTFE Retainers and knob
Latch lid
Documents