missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

DWK Life Sciences Kimble™ Tooled Neck N-51A Borosilicate Glass Vials with Unattached Polyethylene Closure

Tooled neck OPTICLEAR™ vials are excellent for packaging drug products.  Available in a range of capacities (4mL, 12mL, 20mL) in shelf packs of 72 .

$82.50 - $190.00

Specifications

Classification B
Color Clear
Material Borosilicate Glass
Closure Material Polyethylene
Closure Type Open Bottom Stopper
View More Specs

Products 3
Catalog Number Mfr. No. Diameter (Metric) Outer Volume (Metric) Height (Metric) Volume (English) Price Quantity  
Catalog Number Mfr. No. Diameter (Metric) Outer Volume (Metric) Height (Metric) Volume (English) Price Quantity  
03-339-10C
View Documents Promotion Details
DWK Life Sciences
60975L3
21 mm 12 mL 50 mm 3.2 dr.
Pack of 72 Each for $134.47
Case of 6 Pack for $806.82
 
03-339-10E
View Documents Promotion Details
DWK Life Sciences
60975L5
27 mm 20 mL 55 mm 5.4 dr.
Pack of 72 Each for $190.00
Case of 3 Pack for $570.00
 
03-339-10A
View Documents Promotion Details
DWK Life Sciences
60975L1
15 mm 4 mL 45 mm 1 dr.
Pack of 72 Each for $82.50
Case of 12 Pack for $990.00
 
Description

Description

Tooled neck OPTICLEAR™ vials are excellent for packaging drug products.  Available in a range of capacities (4mL, 12mL, 20mL) in shelf packs of 72 .

  • Necks are tooled for an accurate fit with polyethylene closures
  • Vials and closures are supplied in both the pack and case quantities
  • Supplied with closed-bottom, two-piece closures
  • Manufactured from 51 expansion borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class B requirements
  • Options:
    • Capacity 4mL / 1 dram: Height 45mm, OD 15mm, Case of 864
    • Capacity 12mL / 3 drams: Height 50mm, OD 21mm, Case of 432
    • Capacity 20mL / 5 drams: Height 55mm, OD 27mm, Case of 216
    Specifications

    Specifications

    B
    Borosilicate Glass
    Open Bottom Stopper
    Clear
    Polyethylene
    ASTM E438, USP Type I, Type I, Class B
    SDS
    missing translation for 'documents'

    missing translation for 'documents'