DWK Life Sciences Kimble™ Kontes™ Friedrich Condensers
High-quality, water-cooled condensers to meet your vapor- or air-cooling needs
$796.06 - $848.97
Specifications
Material | Borosilicate Glass |
---|---|
Type | Friedrich Condenser |
For Use With (Application) | Provide a long vapor path, good heat transfer and anti-flooding characteristics |
Quantity | Each |
Catalog Number | Mfr. No. | Standard Taper Joint No | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Standard Taper Joint No | Price | Quantity | |||||
K437000-2440
|
DWK Life Sciences
4370002440 |
24/40 |
Each for $796.06
|
|
|||||
K456250-0022
|
DWK Life Sciences
4562500022 |
45/50 |
Each for $848.97
|
|
|||||
Description
Condensers provide three joint sizes to tackle a wide range of tasks.
- Molded inner cold finger
- Hose connection for 3/8 in. ID flexible tubing
24/40 Joint
- Specially designed to provide a long vapor path, good heat transfer and anti-flooding characteristics
- Inclined outer joint on the side and two hose connectors at the top
- Inner drip joint at the bottom
- Side arm is an outer standard taper joint and the bottom is an inner standard taper joint
- Used primarily in reflux mode and Soxhlet extractions
- Water inlet and outlet located ring-sealed inner condensing tube
- Inner drip joint and side outlet for drying tube
Specifications
Borosilicate Glass | |
Friedrich Condenser | |
Provide a long vapor path, good heat transfer and anti-flooding characteristics | |
Each |