missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
DWK Life Sciences Kimble™ Kontes™ Disposable Microtubes
$43.74 - $223.97
Catalog Number | Mfr. No. | Capacity | Color | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity | Color | Price | Quantity | ||||
K749560-1500
|
DWK Life Sciences
7495601500 |
0.05 oz., 1.5mL | Natural |
Case of 500 Each for $223.97
|
|
||||
K749510-1500
|
DWK Life Sciences
7495101500 |
1.5 mL | Blue |
Pack of 100 Each for $84.59
|
|
||||
K749530-1500
|
DWK Life Sciences
7495301500 |
0.5 oz., 15mL | Natural |
Case of 25 Each for $43.74
|
|
||||
Description
- Autoclavable polypropylene
- Selected to give best fit to Pellet Pestles (not included)
- Available colors: blue, natural
- Sizes: 1.5, 15, 50mL
Order Info
Pellet pestle sold separately.
Specifications
Yes | |
Polypropylene |