DWK Life Sciences Kimble™ Kontes™ Full-Length Standard Taper Pennyhead Stopper
Stoppers are used with laboratory glassware such as flasks, mixing cylinders and separatory funnels.
Supplier: DWK Life Sciences 8510002440
Description
- Sizes Standard Taper 10/30 through Standard Taper 19/38 are solid
- Sizes Standard Taper 24/40 and larger are hollow
- Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
Specifications
Pennyhead Stopper | |
Glass | |
24/40 |
Case of 1 | |
Laboratory glassware such as flasks, mixing cylinders and separatory funnels |