DWK Life Sciences Kimble™ KIMAX™ Flasks with Quick-Release Hose Connector
Allow safe attachment of 0.25 in. I.D. (6mm) tubing
$192.15 - $1128.03
Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity | |||||
10-181-7B
|
DWK Life Sciences
27070250 |
250 mL |
Case of 2 Each for $192.15
|
|
|||||
10-181-7C
|
DWK Life Sciences
27070500 |
500 mL |
Case of 8 Each for $1,128.03
|
|
|||||
10-181-7E
|
DWK Life Sciences
270702000 |
2000 mL |
Case for $417.65
|
|
|||||
10-181-7D
|
DWK Life Sciences
270701000 |
1000 mL |
Case for $222.82
|
|
|||||
10-181-7F
|
DWK Life Sciences
270704000 |
4 L |
Case for $589.09
|
|
|||||
Description
Allow safe attachment of 0.25 in. I.D. (6mm) tubing
- Tubing remains permanently attached to hose barb and opposite end attaches to flask tubulation with positive threaded seal
- Proper positioning of connector provides greater flask stability as tubing angled downward has less tendency to tilt flask
- Graduated with approximate volume scale in durable white ceramic enamel
- Tubing and stoppers sold separately
Specifications
Borosilicate Glass |