DWK Life Sciences Kimble™ KIMAX™ Reusable Test Tubes with Beaded Rim and Marking Spot
Marking spot is durable fused-on white enamel
$35.63 - $1677.59
Specifications
Certifications/Compliance | ASTM E982, Type I |
---|---|
Design Type | Plain Beaded Rim |
Material | Borosilicate Glass |
Label Type | Marking Spot |
Catalog Number | Mfr. No. | Capacity (Metric) | Diameter (Metric) | Length (Metric) | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (Metric) | Diameter (Metric) | Length (Metric) | Price | Quantity | |||
14-956A
|
DWK Life Sciences
450421075 |
4 mL | 10 mm | 75 mm |
Pack of 72 Each for $117.09
Case of 10 Pack for $1,170.90
|
|
|||
14-956C
|
DWK Life Sciences
4504213100 |
10 mL | 13 mm | 100 mm |
Pack of 72 Each for $137.74
Case of 10 Pack for $1,377.40
|
|
|||
14-956D
|
DWK Life Sciences
4504215125 |
16 mL | 15 mm | 125 mm |
Pack of 72 Each for $35.63
Case of 10 Pack for $356.30
|
|
|||
14-956E
|
DWK Life Sciences
4504216150 |
24 mL | 16 mm | 150 mm |
Pack of 72 Each for $186.62
Case of 8 Pack for $1,492.96
|
|
|||
14-956F
|
DWK Life Sciences
4504218150 |
28 mL | 18 mm | 150 mm |
Case of 576 Each for $1,677.59
|
|
|||
14-956K
|
DWK Life Sciences
4504225150 |
55 mL | 25 mm | 150 mm |
Case of 288 Each for $1,504.74
|
|
|||
Description
Marking spot is durable fused-on white enamel
- Kimax KG-33 glass
- With sturdy lip
- Uniform wall thickness for maximum heat transfer, chemical resistance
Specifications
ASTM E982, Type I | |
Plain Beaded Rim | |
Borosilicate Glass | |
Marking Spot |