missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Invitrogen™ KDalert™ GAPDH Assay Kit with Manual
KDalert™ GAPDH Assay Kit with Manual
Supplier: Invitrogen™ AM1639M
Description
The Ambion™ KDalert™ GAPDH Assay Kit is for the reliable measure of GAPDH enzyme activity in cultured human, mouse, or rat cells in less than 30 minutes using a microplate fluorometer. The kit includes sufficient reagents for 375 reactions.The KDalert GAPDH Assay Kit is an ideal positive control for transfection optimization experiments and also measures transfection induced cytoxicity. It is designed for use with Ambion™ Silencer™ GAPDH siRNA.
Rapid, Time-Saving Procedure
Use the assay to optimize siRNA transfection by transfecting individual cell samples with a GAPDH siRNA and a negative control siRNA. Two to three days after transfection, simply add the included cell lysis buffer to the cells, incubate for 20 minutes, add the diluted master mix of assay reagents, and read the increase in fluorescence four minutes later using a microplate or standard fluorometer. The assay procedure can be completed in about 30 minutes with minimal sample handling.
One Assay for Two Readouts
Because GAPDH is expressed at relatively constant levels, the assay can also be used to monitor transfection agent induced toxicity. For this analysis, GAPDH enzyme activity from negative control siRNA-transfected cells is compared to that of untreated cells. Reduced GAPDH activity in negative control-transfected cells compared to non-transfected cells is an indication that the transfection-induced cytotoxicity.
Accessory Products:
The KDalert™ Kit is designed for use with Silencer™ GAPDH siRNAs (SKUs #AM4605, AM4633, AM4634, AM4624, AM4632, or AM4631). Additional KDalert™ Lysis Buffer (SKU #AM8790G) is also available separately.
Specifications
KDalert™ GAPDH Assay Kit with Manual, 375 rxns, GAPDH Assays, Fluorescent Detection Method, Chemical substrate, GAPDH (Enzyme) Target, Absorbance, Fluorescence Intensity Technique, Used For RNAi Applications | |
Fluorescence | |
Chemical | |
RNAi, epigenetics and non-coding RNA research, RNAi, siRNA | |
375 Reactions | |
GAPDH (enzyme) |
GAPDH | |
KDalert™ GAPDH Assay Kit | |
375 rxn(s) | |
KDalert lysis buffer can be stored at –20°C or 4°C, solution A, solution B, and solution C should be stored at –20°C, GAPDH enzyme should be stored at or below –70°C | |
For research use only (RUO) |
For Research Use Only. Not for use in diagnostic procedures.