missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Justrite™ Flammables Undercounter Storage Cabinet

Helps to keep work area safely organized

Manufacturer:  Justrite™ JMC892300

Catalog No. 29-635-9870



Includes: One adjustable shelf

Description

Description

  • Double-wall construction provides 1-1/2 in. of insulating air space to help prevent the spread of flames High-visibility Haz-Alert™ reflective warning labels offer added safety during power outages or fires
  • Dual vents with flame arresters at top and opposite bottom allow hazardous vapors to be piped away Easy-closing, self-latching doors facilitate fire protection of the cabinet
  • Self latch engages critical three-point latching system for protection under fire conditions
  • Two self-latching manual doors
  • Fusible links hold self-closing doors wide open, flush with front of cabinet for easy access and resupply; automatically release and slam doors shut at 74°C (165°F)

  • Concealed self-close mechanism provides optimum storage space and obstruction-free access to contents
  • SpillSlope safety shelves direct incidental spills to the back and bottom of leakproof sump
  • Exterior side panel has built-in grounding connector for easy grounding
  • Finished inside and out with durable lead-free powder paint
  • Dual vents with flame arresters in back walls
  • Kickplate (3.5 × 2.5 in. [8.9 × 6.4cm]) mounts flush with other cabinetry
  • Leveling feet adjust to 1.13 in. (2.9cm) for alignment on uneven floors
  • Capacity: 22 gal. (3L)
  • Backed by an exclusive 10-year warranty
  • Meet OSHA and NFPA requirements

Order Info

Dimensions: 22L × 35W × 35 in.H (56 × 89 × 89cm).

Certifications

FM approved.

Compliance

Meets OSHA and NFPA code 30 requirements.

Specifications

Specifications

22 gal.
18.75 in.
47.6cm
Manual-close
27.37 in.
69.5cm
2
Epoxy, Polyester
189.59 lb.
31.75 in.
80.6cm
22 in.
Steel (CR)
FM, NFPA, OSHA
55.9cm
18.75 x 31.75 x 27.37 in. (47.6 x 80.6 x 69.5cm)
Flammables
88.9cm
One adjustable shelves and mounting hardware, U-Loc™ handles with two keys, Haz-Alert™ reflective trilingual exterior labeling and Galvanized Steel SpillSlope™ shelves that direct all spills to the 5.1
1
Undercounter safety cabinet
86kg
88.9cm
Yellow
35 in.
35 in.
Documents