Learn More
Isonicotinamide, 99%, Thermo Scientific Chemicals
$62.88 - $368.07
Chemical Identifiers
CAS | 1453-82-3 |
---|---|
Molecular Formula | C6H6N2O |
Molecular Weight (g/mol) | 122.127 |
MDL Number | MFCD00006432 |
InChI Key | VFQXVTODMYMSMJ-UHFFFAOYSA-N |
Synonym | isonicotinamide, 4-pyridinecarboxamide, 4-carbamoylpyridine, isonicotinic acid amide, isonicotineamide, gamma-pyridinecarboxamide, pyridine-4-carboxylic acid amide, unii-4h3bh6yx9q, .gamma.-pyridinecarboxamide, 4h3bh6yx9q |
PubChem CID | 15074 |
ChEBI | CHEBI:6031 |
IUPAC Name | pyridine-4-carboxamide |
SMILES | C1=CN=CC=C1C(=O)N |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1819018
|
Thermo Scientific Chemicals
A1819018 |
50 g |
Each for $62.88
|
|
|||||
AAA1819022
|
Thermo Scientific Chemicals
A1819022 |
100 g |
Each for $101.81
|
|
|||||
AAA1819030
|
Thermo Scientific Chemicals
A1819030 |
250 g |
Each for $218.12
|
|
|||||
AAA1819036
|
Thermo Scientific Chemicals
A1819036 |
500 g |
Each for $368.07
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1453-82-3 | |
122.127 | |
VFQXVTODMYMSMJ-UHFFFAOYSA-N | |
15074 | |
pyridine-4-carboxamide |
C6H6N2O | |
MFCD00006432 | |
isonicotinamide, 4-pyridinecarboxamide, 4-carbamoylpyridine, isonicotinic acid amide, isonicotineamide, gamma-pyridinecarboxamide, pyridine-4-carboxylic acid amide, unii-4h3bh6yx9q, .gamma.-pyridinecarboxamide, 4h3bh6yx9q | |
CHEBI:6031 | |
C1=CN=CC=C1C(=O)N |
Specifications
1453-82-3 | |
C6H6N2O | |
50 g | |
isonicotinamide, 4-pyridinecarboxamide, 4-carbamoylpyridine, isonicotinic acid amide, isonicotineamide, gamma-pyridinecarboxamide, pyridine-4-carboxylic acid amide, unii-4h3bh6yx9q, .gamma.-pyridinecarboxamide, 4h3bh6yx9q | |
C1=CN=CC=C1C(=O)N | |
122.127 | |
CHEBI:6031 | |
99% |
155°C to 158°C | |
MFCD00006432 | |
2173 | |
VFQXVTODMYMSMJ-UHFFFAOYSA-N | |
pyridine-4-carboxamide | |
15074 | |
122.13 | |
Isonicotinamide |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 215-926-2
RTECSNumber : NR9500000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only