missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Iron(II) sulfide, tech., Thermo Scientific Chemicals

$56.66 - $668.18

Chemical Identifiers

CAS 1317-37-9
Molecular Formula FeS
Molecular Weight (g/mol) 87.91
MDL Number MFCD00011013
InChI Key GNVXPFBEZCSHQZ-UHFFFAOYSA-N
Synonym ferrous sulfide, iron ii sulfide, iron sulfide, troilite, iron sulfuret, iron sulphide, iron monosulfide, iron protosulfide, ferrous monosulfide
PubChem CID 14828
SMILES [S--].[Fe++]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1556930
missing translation for 'viewDocuments' Promotion Details
Thermo Scientific Chemicals
A1556930
250 g
Each for $56.66
 
AAA155690B
missing translation for 'viewDocuments' Promotion Details
Thermo Scientific Chemicals
A155690B
1000 g
Each for $161.92
 
AAA155690I
missing translation for 'viewDocuments' Promotion Details
Thermo Scientific Chemicals
A155690I
5000 g
Each for $668.18
 
Description

Description

Iron(II) sulfide is used as a pigment in hair dyes and ceramics, glasses and bottles, and paint, Also used in lubricants and anodes, as a hydrogen sulfide source, and to treat exhaust gases. And it is also used as resulphurizing agent in the production of carbon, alloy and stainless types of free cutting steels. It acts as an alloying agent to improve machine ability of steel castings which are used to manufacture various steel components.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Iron(II) sulfide is used as a pigment in hair dyes and ceramics, glasses and bottles, and paint, Also used in lubricants and anodes, as a hydrogen sulfide source, and to treat exhaust gases. And it is also used as resulphurizing agent in the production of carbon, alloy and stainless types of free cutting steels. It acts as an alloying agent to improve machine ability of steel castings which are used to manufacture various steel components.

Solubility
Soluble in water(0.0062g/L ), in acids with evolution of hydrogen sulfide. Insoluble in nitric acid.

Notes
Moisture Sensitive. Store in a cool, dry, well-ventilated area away from incompatible substances such as oxidizing agent, acids, air, water/ moisture.
Specifications

Chemical Identifiers

1317-37-9
87.91
GNVXPFBEZCSHQZ-UHFFFAOYSA-N
14828
FeS
MFCD00011013
ferrous sulfide, iron ii sulfide, iron sulfide, troilite, iron sulfuret, iron sulphide, iron monosulfide, iron protosulfide, ferrous monosulfide
[S--].[Fe++]

Specifications

1195°C
250 g
MFCD00011013
ferrous sulfide, iron ii sulfide, iron sulfide, troilite, iron sulfuret, iron sulphide, iron monosulfide, iron protosulfide, ferrous monosulfide
GNVXPFBEZCSHQZ-UHFFFAOYSA-N
λ²-iron(2+) sulfanediide
14828
Technical
Air and moisture sensitive
Iron(II) sulfide
1317-37-9
FeS
14,4058
Soluble in water(0.0062g/L ),in acids with evolution of hydrogen sulfide. Insoluble in nitric acid.
[S--].[Fe++]
87.91
87.91
Odorless
4.74
Safety and Handling

Safety and Handling

EINECSNumber : 215-268-6

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Argon

SDS
Documents

Documents

RUO – Research Use Only