Learn More
Iron(II) sulfate heptahydrate, ACS, 99+%, Thermo Scientific Chemicals
Air Sensitive
$81.08 - $259.12
Chemical Identifiers
CAS | 7782-63-0 |
---|---|
Molecular Formula | FeH14O11S |
Molecular Weight (g/mol) | 278.01 |
MDL Number | MFCD00149719 |
InChI Key | SURQXAFEQWPFPV-UHFFFAOYSA-L |
Synonym | iron ii sulfate heptahydrate, ferrous sulfate heptahydrate, presfersul, melanterite mineral, iron sulfate heptahydrate, iron 2+ sulfate heptahydrate, fesofor, fesotyme, haemofort, ironate |
PubChem CID | 62662 |
ChEBI | CHEBI:75836 |
SMILES | O.O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O |
Description
Iron(II) sulfate heptahydrate is used as a precursor to prepare other iron compounds such as a lawn conditioner and a mordant for wool dyeing. It is actively used in the manufacture of ink including iron gall ink. As a reducing agent, it participates in the reduction of chromate in cement. It is also used in industrial water treatment plants to remove phosphate. It is also used in the gold refining process to precipitate metallic gold. Woodworkers use aqueous solutions of ferrous sulfate to color maple wood a silvery hue. It finds application in the treatment of iron chlorosis, which arises due to deficiency of iron in horticulture.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7782-63-0 | |
278.01 | |
SURQXAFEQWPFPV-UHFFFAOYSA-L | |
62662 | |
O.O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O |
FeH14O11S | |
MFCD00149719 | |
iron ii sulfate heptahydrate, ferrous sulfate heptahydrate, presfersul, melanterite mineral, iron sulfate heptahydrate, iron 2+ sulfate heptahydrate, fesofor, fesotyme, haemofort, ironate | |
CHEBI:75836 |
Specifications
64°C-6H2O | |
Green | |
250 g | |
FeH14O11S | |
iron ii sulfate heptahydrate, ferrous sulfate heptahydrate, presfersul, melanterite mineral, iron sulfate heptahydrate, iron 2+ sulfate heptahydrate, fesofor, fesotyme, haemofort, ironate | |
SURQXAFEQWPFPV-UHFFFAOYSA-L | |
iron(2+);sulfate;heptahydrate | |
62662 | |
278.02 (151.91 Anhydrous) | |
Air Sensitive | |
1.897 g/cm3 |
7782-63-0 | |
Crystalline | |
>99% | |
MFCD00149719 | |
Soluble in water. Insoluble in alcohol. | |
O.O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O | |
278.01 | |
CHEBI:75836 | |
ACS | |
Green crystalline | |
Iron(II) sulfate heptahydrate, For ACS analysis |
Safety and Handling
P260-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P314-P330-P332+P313-P362-P501c
H302-H315-H319-H336-H373
EINECSNumber : 231-753-5
RTECSNumber : NO8510000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only