missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Instrument Check Standard 6 For Ion Chromatography, SPEX CertiPrep™

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Manufacturer:  SPEX CertiPrep Group LLC ICMIX6100

Catalog No. ICMIX6-100



Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

Description

Description

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
HPO4-2150μg/mL
SO4-2150μg/mL
Br-100μg/mL
Cl-50μg/mL
NO2-N25μg/mL
NO3-N25μg/mL
F-20μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Specifications

Instrument Check Standard 6
Triple-leached LDPE bottle
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
125mL
Components in water:
Br-    •100μg/mL
Cl-    •50μg/mL
F-    •20μg/mL
HPO4-2    •150μg/mL
NO3--N    •25μg/mL
NO2--N    •25μg/mL
SO4-2    •150μg/mL
With Certificate of Analysis
H2O
AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Documents
Promotions

Promotions

Product intended for laboratory use only