missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Invitrogen™ Total Exosome Isolation Reagent (from cell culture media)
Total Exosome Isolation Reagent (from cell culture media) enables fast and efficient enrichment of intact exosomes from cell culture media samples.
Supplier: Invitrogen™ 4478359
Description
Total Exosome Isolation Reagent (from cell culture media) enables fast and efficient enrichment of intact exosomes from cell culture media samples.
- Maximize recovery of intact exosomes for any type of downstream application
- Maximize flexibility—can be scaled up or down depending on the sample size
- Enables simple and reliable concentration of intact exosomes from cell media samples
Order Info
Shipping Condition: Room Temperature
Specifications
Store at 2°C to 8°C. | |
Exosomes | |
Room Temperature |
Total Exosome Isolation | |
Cell Culture Media |
For Research Use Only. Not for use in diagnostic procedures.