Learn More
Invitrogen™ SuperScript™ II Reverse Transcriptase
Invitrogen SuperScript II Reverse Transcriptase is a genetically engineered MMLV reverse transcriptase (RT) with reduced RNase H activity and increased thermal stability compared to wild-type MMLV RT.
$162.00 - $2100.00
Specifications
Format | Stand-alone Enzyme |
---|---|
GC-Rich PCR Performance | High |
Reaction Speed | 50 min. |
Technique | Reverse Transcription |
Optimal Reaction Temperature | 42°C |
Description
Invitrogen SuperScript II Reverse Transcriptase is a genetically engineered MMLV reverse transcriptase (RT) with reduced RNase H activity and increased thermal stability compared to wild-type MMLV RT. Mutations in the RNase H domain of the enzyme eliminate degradation of the RNA during first-strand cDNA synthesis, which results in higher yields of full-length cDNA.
SuperScript reverse transcriptases are the most highly trusted and widely used RTs with over 50,000 citations, reviews, and publications to date. The enzyme is purified to near homogeneity from E. coli containing the modified pol gene of Moloney Murine Leukemia Virus.
Features of SuperScript II Reverse Transcriptase include:
• High cDNA yields and more full-length product
• Generation of cDNA up to 12.3 kb
• First-strand cDNA synthesis at higher temperatures than conventional MMLV RT
Note: The latest member of the SuperScript RT family, SuperScript IV Reverse Transcriptase (Cat. No. 18090010), features enhanced thermostability, processivity, yields, and performance with any RNA samples, including those of suboptimal purity or integrity.
Order Info
Shipping Condition: Wet ice
Specifications
Stand-alone Enzyme | |
50 min. | |
42°C | |
Reduced | |
200 U/μL | |
Separate components | |
Up to 12.3 kb |
High | |
Reverse Transcription | |
SuperScript II | |
Wet Ice | |
First-Strand cDNA | |
Reverse Transcription | |
RNA |
For Research Use Only. Not for use in diagnostic procedures.