missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ Oligo(dT)12-18 Primer DFS Item

Oligo(dT)12-18 Primer is suitable for use in first-strand cDNA synthesis with reverse transcriptase.

Supplier:  Invitrogen™ 18418012

Catalog No. 18418012


Description

Description

Oligo(dT)12-18 Primer is suitable for use in first-strand cDNA synthesis with reverse transcriptase. The primer hybridizes to the poly(A) tail of mRNA. It is phosphorylated on the 5´ end to facilitate cloning of cDNA.

Performance and quality testing:
Performance is evaluated in a first-strand cDNA synthesis reaction

Order Info

Shipping Conditions: Approved for shipment on Wet or Dry Ice

Specifications

Specifications

Primer
Phosphate
Gel-purified, Desalted
0.5 μg/μL
50 μL
Liquid
• Oligo(dT)12-18 Primer in DEPC-treated water (50 μg at 0.5 μg/μL)

Store at –20°C.
Guaranteed stable for 6 months when properly stored.
5'd PO4 [(T)12-18] 3'
Approved for shipment on Wet or Dry Ice
Oligo dT
First-strand cDNA Synthesis
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.