missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ Lipofectamine™ 2000 Transfection Reagent DFS Item

Versatile transfection reagent that has been shown to effectively transfect the widest variety of adherent and suspension cell lines

$880.00 - $7,010.00

Specifications

For Use With (Application) Plasmid DNA, RNAi Plasmids (shRNA, miR), Synthetic siRNA
Storage Requirements Store at 2° to 8°C. Do not freeze.
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
11668500 View Documents Invitrogen™
LS11668500
15mL Each for $7,010.00
11668019 View Documents Invitrogen™
LS11668019
1.5mL Each for $880.00
Description

Description

Lipofectamine™ 2000 Transfection Reagent is versatile transfection reagent that has been shown to effectively transfect widest variety of adherent and suspension cell lines. Researchers use Lipofectamine™ 2000 Reagent for siRNA- and shRNA-based gene knockdown experiments, as well as for gene expression studies. Lipofectamine 2000 Transfection Reagent is best transfection reagent available from Life Technologies for the co-transfection of siRNA and plasmid DNA.

  • Exceptional transfection efficiency in the broadest range of cell lines and the highest levels of recombinant protein expression
  • Superior performance for co-transfection of siRNA and plasmid DNA
  • Proven efficacy in the presence of serum, eliminates the need to change media following transfection
  • Reliable performance for high-throughput applications
  • The best choice for establishing stable cell lines

Order Info

Sustainable packaging. Approved for shipment at Room Temperature or on Wet Ice.

Specifications

Specifications

Plasmid DNA, RNAi Plasmids (shRNA, miR), Synthetic siRNA
Store at 2° to 8°C. Do not freeze.

For Research Use Only. Not for use in diagnostic procedures.