missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ dNTP Set (100 mM) Solution DFS Item

Ready-to-use solution

$304.00 - $1,726.00

Specifications

Concentration 100mM
Conjugate Unlabeled
Storage Requirements Store at -30° to -10°C.
View More Specs

Products 2 Description

Description

The 100mM dNTP Set consists of four deoxynucleotides (dATP, dCTP, dGTP, dTTP), each at a concentration of 100mM. The deoxynucleotides are suitable for use in polymerase chain reaction (PCR), sequencing, fill-in, nick translation, cDNA synthesis, and TdT tailing reactions.

ChIP-on-Chip, Chromatin Biology, PCR, PCR and Real-Time PCR, RNAi, Epigenetics and Non-Coding RNA Research

Order Info

Approved for shipment on Wet or Dry Ice

Specifications

Specifications

100mM
Unlabeled
Store at -30° to -10°C.

For Research Use Only. Not for use in diagnostic procedures.