missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ dNTP Mix (10 mM) DFS Item

Mix of four nucleotides

$32.74 - $217.00

Specifications

Concentration 10mM
Storage Requirements Store at -20°C. Guaranteed stable for 6 months when properly stored.
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
18427088 View Documents Invitrogen™
LS18427088
1mL Each for $217.00
18427013 View Documents Invitrogen™
LS18427013
100μL Each for $32.74
Description

Description

The 10mM dNTPmix consists dATP, dCTP, dGTP and dTTP (2'-deoxynucleoside 5'-triphosphates), each at a concentration of 10mM, in a solution of 0.6mM Tris-HCl (pH 7.5). The 10mM dNTPmix is suitable for use in PCR, sequencing, fill-in, nick translation, cDNA synthesis, TdT-tailing reactions, and dilution of radiolabeled dNTPs.

PCR, PCR and Real-Time PCR

Order Info

Approved for shipment on Wet or Dry Ice

Specifications

Specifications

10mM
Store at -20°C. Guaranteed stable for 6 months when properly stored.

For Research Use Only. Not for use in diagnostic procedures.