Invitrogen™ Countess™ Cell Counting Chamber Slides DFS Item
Designed for use with Countess Automated Cell Counter
$119.00 - $9,530.00
Specifications
For Use With (Equipment) Cell Counting Chamber
View More Specs
Includes: Sildes and 20 × 1mL vials of trypan blue (0.4%)
Products 5
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
C10228 View Documents Invitrogen™
LSC10228
50 slides Each for $119.00
C10312 View Documents Invitrogen™
LSC10312
500 slides Each for $1,142.00
C10314 View Documents Invitrogen™
LSC10314
2500 slides Each for $5,210.00
C10313 View Documents Invitrogen™
LSC10313
1250 slides Each for $2,760.00
C10315 View Documents Invitrogen™
LSC10315
5000 slides Each for $9,530.00
Description
Description
  • Presterilized and individually packaged
  • Features two separate, enclosed chambers (75 × 25 × 1.8mm; chamber depth = 100μm) used to count one sample in duplicate or to count two different samples
  • Cell counting occurs in central location of the counting chamber
  • Sample Loading Volume: 10μL
Cell Counting, Viability and Cryopreservation
Specifications
Specifications
Cell Counting Chamber