missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Invitrogen™ Gateway™ BP Clonase™ II Enzyme mix
Catalyzes in vitro recombination of PCR products to generate entry clones
$588.00 - $2560.00
Specifications
Content And Storage | Gateway™ BP Clonase™ II enzyme mix includes proteinase K solution (2 μg/μl) and a positive control vector. Store at -20°C or -80°C. Guaranteed stable for 6 months when properly stored. |
---|---|
Shipping Condition | Dry Ice |
Enzyme | BP Clonase |
Compatible Buffer | Enzyme Buffer |
Product Type | BP Clonase Enzyme Mix |
Includes: Proteinase K solution (2μg/μL) and a positive control vector
Description
- Catalyzes the in vitro recombination of PCR products or subcloning DNA segments from clones (containing attB sites) and a donor vector (containing attP sites) to generate entry clones
- Contains enzymes and buffer in a single mix to enable convenient ten-microliter reaction set up with fewer pipetting steps
- Guaranteed stable for 6 months when properly stored at -20°C to -80°C
Order Info
Shipping Conditions: Dry ice
Specifications
Gateway™ BP Clonase™ II enzyme mix includes proteinase K solution (2 μg/μl) and a positive control vector. Store at -20°C or -80°C. Guaranteed stable for 6 months when properly stored. | |
BP Clonase | |
BP Clonase Enzyme Mix |
Dry Ice | |
Enzyme Buffer | |
Clonase™, Gateway™ |
For Research Use Only. Not for use in diagnostic procedures.