Invitrogen™ iBright FL1000 Imager DFS Item
iBright FL1000 Imager is a powerful, easy-to-use western blot and gel documentation system, providing sensitive, streamlined, multimode image capture.
Manufacturer:  Invitrogen™ LSA32752
Catalog No. A32752
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
The Invitrogen™ iBright™ FL1000 Imager is a powerful, easy-to-use western blot and gel documentation system. The iBright FL1000 Imager provides sensitive, streamlined, multimode image capture. The iBright FL1000 connects to the user through a large capacitive touch screen interface and intelligently designed software to provide stunningly easy western blot and gel imaging.
The iBright FL1000 Imager revolutionizes image acquisition of western blots and gels. The compact benchtop instrument is equipped with five fluorescent channels: three in the visible fluorescence range (RGB) and two in the near infrared (NIR) fluorescence range, which enables compatibility with a wide range of fluorescence dyes.  Multiplex up to four channels simultaneously in a single western blot.
The iBright FL1000 provides fast, accurate, chemiluminescent detection using an advanced 9.1 MP CCD camera to capture chemiluminescent images with high sensitivity and dynamic range. The incorporated advanced CCD technology results in greater to or equal sensitivity to X-ray film and provides enhanced dynamic range and signal linearity.
Capture stained nucleic acid gels (e.g. ethidium bromide, SYBR green, SYBR safe etc.) using the integrated environmentally friendly long-life green LED-based transilluminator, which provides a light source without harmful UV rays or mercury waste from UV transilluminator bulbs.
The built-in 12.1 inch LCD touch screen features multi-touch technology that enables pinch-and-zoom, pan-to-move images, and swipe-to-switch between channels and overlays. Acquired images can be easily exported through USB ports, local network drive connection, or Wi-Fi (optional).
The iBright FL1000 Imager is Thermo Fisher Cloud-enabled, providing secure storage and advanced data analysis. Thermo Fisher Cloud is web browser-based software tools that are flexible, fast and easy to use. These software tools are compatible with Mac or PC.
Warranty
One year
Specifications
Specifications
9.1 MP CCD
iBright Imager
Western blotting and Gel documentation
FL1000
Detection mode: Chemiluminescence, RGB Fluorescence, near-IR fluorescence, visible, nucleic acid stains
12.1 inch LCD touch screen
Thermo Fisher Cloud
Documents
For research use only. Not for use in diagnostic procedures.