Learn More
Thermo Scientific™ Hypersil™ BDS C8 HPLC Columns
Use as a robust, general-purpose column for QC/QA applications. They are high-quality, base-deactivated, fully endcapped, have similar C18 selectivity but slightly less retentive.
$1267.95 - $1751.28
Specifications
Pore Size | 130 Å |
---|---|
Surface Area | 170 m²/g |
Temperature | 60°C |
USP Type | L7 |
Stationary Phase | C8 |
Catalog Number | Mfr. No. | Particle Size | Diameter (Metric) | Length (Metric) | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Particle Size | Diameter (Metric) | Length (Metric) | Price | Quantity | |||
03-150-749
|
Thermo Scientific™
28205053030 |
5 μm | 3 mm | 50 mm |
Each for $1,324.42
|
|
|||
28205054630
|
Thermo Scientific™
28205054630 |
5 μm | 4.6 mm | 50 mm |
Each for $1,267.95
|
|
|||
28-205-102130
|
Thermo Scientific™
28205102130 |
5 μm | 2.1 mm | 100 mm |
Each for $1,684.79
|
|
|||
28205104630
|
Thermo Scientific™
28205104630 |
5 μm | 4.6 mm | 100 mm |
Each for $1,385.70
|
|
|||
28-205-154030
|
Thermo Scientific™
28205154030 |
5 μm | 4 mm | 150 mm |
Each for $1,700.81
|
|
|||
03-150-756
|
Thermo Scientific™
28205154630 |
5 μm | 4.6 mm | 150 mm |
Each for $1,509.95
|
|
|||
28205-254630
|
Thermo Scientific™
28205254630 |
5 μm | 4.6 mm | 250 mm |
Each for $1,744.75
|
|
|||
28-203-104630
|
Thermo Scientific™
28203104630 |
3 μm | 4.6 mm | 100 mm |
Each for $1,751.28
|
|
|||
28203054630
|
Thermo Scientific™
28203054630 |
3 μm | 4.6 mm | 50 mm |
Each for $1,395.53
|
|
|||
28203052130
|
Thermo Scientific™
28203052130 |
3 μm | 2.1 mm | 50 mm |
Each for $1,470.11
|
|
|||
Description
For a high-quality, base-deactivated, fully endcapped phase with similar selectivity to C18 but slightly less retention, use Thermo ScientificTM HypersilTM BDS C8 HPLC Columns. These economical columns feature reduced silanol activity and provide exceptional reliability for QA/QC labs. Built on the renowned Hypersil silica backbone, the base-deactivated silica is one of the most popular packing materials available.
For more challenging separations that require improved chromatographic performance, use Thermo ScientificTM Hypersil GOLDTM C8 columns as an alternative.
- Excellent choice for acids, bases and neutral compounds
- Less retentive than C18 for faster analyses
- Robust columns with long column lifetimes
- Very reliable for routine assays
Ordering Information:
Other column dimensions are available. Please call Customer Service for more information.
Specifications
130 Å | |
60°C | |
C8 | |
Reversed Phase | |
Reversed Phase |
170 m²/g | |
L7 | |
7% | |
Yes |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.