missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ HyPerforma™ 50 to 500L Tank Liners

Thermo Scientific™ HyPerforma™ 50 to 500L Tank Liners are available with or without bottom drain and are provided sterile for handling fluids in bioproduction applications.

$61.23 - $442.14


Products 7 Description

Description

Thermo Scientific™ HyPerforma™ 50 to 500L Tank Liners are available with or without bottom drain and are provided sterile for handling fluids in bioproduction applications.

Tank Liner Key Features:

  • Available in sizes that fit into industry standard cylindrical tanks and in Thermo Scientific HyPerforma Drums (SV3011X, SV50076, and SV50517 series)
  • Gusset design allows for a close fit to the tank
  • Chamber constructed of Thermo Scientific CX3-9 film, which provides the same contact surface as other Thermo Scientific BPCs
  • Supplied gamma irradiated
  • Customization options are available by contacting one of our Thermo Scientific Representatives today

Standard Tank Liners for your liquid applications available in-stock in the following configurations:

SH30647.01 to .03:

  • 50 to 200L Tank Liner
  • Support Container Needed: Design to fit into HyPerforma Flat Bottom PE Drums (SV50076 series)
  • Quantity: Each = 1 Liner

SH30647.04 to .07:

  • 50 to 500L Tank Liner
  • Support Container Needed: Designed to fit into Industry Standard Cylindrical Tanks (SV3011X series)
  • Quantity: Each = 1 Liner

SH30646 Series:

  • 50 to 200L Tank Liner with One Bottom Drain
  • Bottom Lineset 1: 72in. (182.9cm) of tubing leading to a Quick Disconnect (MPX series) Insert (male) connector
  • Support Container Needed: Designed to fit into HyPerforma Conical Bottom PE Drums (SV50517 series)

Ordering Information:

Supplied double polybagged and irradiated with Certificate of Analysis.

Recommended for:

  • Hydrations of powdered media and buffers
  • Pooling of non-sterile solutions and fluids
  • Non-aseptic collection of waste liquids from separation and purification systems

To order, use the part numbers listed in the above table.

Component Detail

SH30646 Series:

  • Quick Disconnect Insert (male): 1/2in. (12.7mm) HB, with Female Cap (MPX series); Polycarbonate
  • Tubing: 1/2in. (12.7mm) ID x 1/8in. (3.2mm) Wall Thickness; C-Flex™ (R70-082)
For more detailed information regarding the above product series, go to the Resource Library and select the Component Information Summary (CIS) sheet.