missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing
Hydroxylamine Hydrochloride, 20% (w/v), Ricca Chemical
Chemical Identifiers
CAS |
5470-11-1 |
Molecular Formula |
ClH4NO |
Molecular Weight (g/mol) |
69.49 |
MDL Number |
MFCD00051089 |
InChI Key |
WTDHULULXKLSOZ-UHFFFAOYSA-N |
Synonym |
hydroxylamine hydrochloride, hydroxylamine hcl, hydroxylammonium chloride, oxammonium hydrochloride, hydroxylamine, hydrochloride, hydroxyamine hydrochloride, hydroxylamine chloride 1:1, hydroxylamine, hydrochloride 1:1, hydroxyamine hcl, oxammonium hci |
PubChem CID |
443297 |
ChEBI |
CHEBI:5807 |
IUPAC Name |
hydroxylamine hydrochloride |
SMILES |
Cl.NO |
View More Specs
Products
1
Description
Specifications
Chemical Identifiers
|
5470-11-1 |
|
69.49 |
|
WTDHULULXKLSOZ-UHFFFAOYSA-N |
|
443297 |
|
hydroxylamine hydrochloride |
|
ClH4NO |
|
MFCD00051089 |
|
hydroxylamine hydrochloride, hydroxylamine hcl, hydroxylammonium chloride, oxammonium hydrochloride, hydroxylamine, hydrochloride, hydroxyamine hydrochloride, hydroxylamine chloride 1:1, hydroxylamine, hydrochloride 1:1, hydroxyamine hcl, oxammonium hci |
|
CHEBI:5807 |
|
Cl.NO |
Specifications
|
Colorless |
|
82.0,16.34 |
|
ClH4NO |
|
hydroxylamine hydrochloride, hydroxylamine hcl, hydroxylammonium chloride, oxammonium hydrochloride, hydroxylamine, hydrochloride, hydroxyamine hydrochloride, hydroxylamine chloride 1:1, hydroxylamine, hydrochloride 1:1, hydroxyamine hcl, oxammonium hci |
|
Cl.NO |
|
69.49 |
|
CHEBI:5807 |
|
Aqueous Solution |
|
Liquid |
|
Hydroxylamine Hydrochloride |
|
5470-11-1,7732-18-5 |
|
85.0,17.01 |
|
MFCD00051089 |
|
WTDHULULXKLSOZ-UHFFFAOYSA-N |
|
hydroxylamine hydrochloride |
|
443297 |
|
Natural Poly Bottle |
|
120 mL |
|
20% (w/v) |
SDS
missing translation for 'documents'