missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
HYDRANAL™ - Water Standard 0.1, Standard for Karl Fischer titration (water content 0.1 mg/g = 0.01%), verified against NIST SRM 2890 & NMIJ CRM 4222, Honeywell™ Fluka™
$227.80 - $227.80
Chemical Identifiers
CAS | 95-47-6 |
---|---|
Molecular Formula | C8H10 |
Molecular Weight (g/mol) | 106.17 |
MDL Number | MFCD00008519 |
InChI Key | CTQNGGLPUBDAKN-UHFFFAOYSA-N |
IUPAC Name | 1,2-xylene |
SMILES | CC1=CC=CC=C1C |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
60050008
|
Honeywell-Hydranal
348476X40ML |
1 Kit |
Each for $227.80
Case of 6 Each for $1,366.80
|
|
|||||
Chemical Identifiers
95-47-6 | |
106.17 | |
CTQNGGLPUBDAKN-UHFFFAOYSA-N | |
CC1=CC=CC=C1C |
C8H10 | |
MFCD00008519 | |
1,2-xylene |
Specifications
23 − 27°C | |
0.86Â g/mLÂ at 20Â °C | |
≤100.0000% | |
Ampoule | |
MFCD00008519 | |
CTQNGGLPUBDAKN-UHFFFAOYSA-N | |
1,2-xylene | |
106.17 | |
HYDRANAL™ - Water Standard 0.1, Standard for Karl Fischer titration (water content 0.1 mg/g = 0.01%), exact value on report of analysis |
1 Kit | |
95-47-6,7732-18-5 | |
136°C to 145°C | |
C8H10 | |
UN1307 | |
CC1=CC=CC=C1C | |
106.17 | |
Karl Fischer |
Safety and Handling
ShelfLife : 720 days from date of manufacture
DOTInformation : Transport Hazard Class: 3; Packing Group:3: III; Proper Shipping Name: Xylenes
Recommended Storage : 2° to 8°C