missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

HYDRANAL™ - Water Standard 0.1 PC, Standard for Karl Fischer titration (water content 0.1 mg/g = 0.01%), verified against NIST SRM 2890 & NMIJ CRM 4222, Honeywell Fluka™

Manufacturer:  Honeywell Chemicals 3444640ML

Catalog No. 60050003



Specifications

Specifications

HYDRANAL™ - Water Standard 0.1 PC
135°C
10 x 4mL
108-32-7
Ampoule
RUOJZAUFBMNUDX-UHFFFAOYSA-N
4-methyl-1,3-dioxolan-2-one
7924
1.205 g/cm3
242°C
Standard for Karl Fischer titration (water content 0.1 mg/g = 0.01%), exact value on report of analysis
≤100.0000%
C4H6O3
CC1COC(=O)O1
102.089
Karl Fischer

Chemical Identifiers

108-32-7
102.089
7924
CC1COC(=O)O1
C4H6O3
RUOJZAUFBMNUDX-UHFFFAOYSA-N
4-methyl-1,3-dioxolan-2-one
Safety and Handling

Safety and Handling

ShelfLife : 1800 days from date of manufacture

Recommended Storage : Room Temp

Documents