missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

HYDRANAL™ - Coulomat CG, Reagent for coulometric KF titration (catholyte solution), Honeywell Fluka™

$121.94 - $991.20

Specifications

Chemical Name or Material HYDRANAL™-Coulomat CG
Quantity 10 x 5mL ampules
Flash Point 14°C (57.2°F)
UN Number UN1230
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
6003350 View Documents Honeywell Chemicals
348406X50ML
1 kit Each for $991.20
 
6000649 View Documents Honeywell Chemicals
3484050ML
10 x 5mL ampules Each for $121.94
 
Specifications

Specifications

HYDRANAL™-Coulomat CG
10 x 5mL ampules
14°C (57.2°F)
UN1230