missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

HYDRANAL™ - Composite 5 K, Reagent for volumetric one-component Karl Fischer titration in aldehydes and ketones (methanol free), Honeywell Fluka™

$124.48 - $2,484.00

Specifications

Chemical Name or Material HYDRANAL™-Composite 5 K
Density 1.15g/mL
Flash Point 90°C (194°F)
View More Specs

Products 6
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
6002569 View Documents Honeywell International
348161LUS
1L Each for $201.08
6002570 View Documents Honeywell International
3481625LUS
2.5L Each for $446.17
6002571 View Documents Honeywell International
34816500MLUS
500mL Each for $124.48
60-046-466 View Documents Honeywell International
348164X25LUS
4 x 2.5L Case of 4 Each for $2,484.00
60-046-468 View Documents Honeywell International
348166X1LUS
6 x 1L Case of 6 Each for $1,696.91
60-046-469 View Documents Honeywell International
348166X500MLUS
6 x 500mL Case of 6 Each for $1,043.23
Specifications

Specifications

HYDRANAL™-Composite 5 K
1.15g/mL
90°C (194°F)