missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

HYDRANAL™ - Composite 5, Reagent for volumetric one-component Karl Fischer titration (methanol free), Honeywell Fluka™

$92.58 - $316.54

Specifications

Chemical Name or Material HYDRANAL™-Composite 5
Density 1.15g/mL
Flash Point 90°C (194°F)
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
6002563 View Documents Honeywell International
348056X1LUS
1L Each for $145.91
6002564 View Documents Honeywell International
348054X25LUS
2.5L Each for $316.54
6002565 View Documents Honeywell International
348056X500MLUS
500mL Each for $92.58
Specifications

Specifications

HYDRANAL™-Composite 5
1.15g/mL
90°C (194°F)