missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

HYDRANAL™ - Composite 2, Reagent for volumetric one-component Karl Fischer titration (methanol free), Honeywell Fluka™

$92.58 - $1,763.60

Specifications

Chemical Name or Material HYDRANAL™ - Composite 2
Density ∼1.11 g/mL
Flash Point 103°C
View More Specs

Products 6 Specifications

Specifications

HYDRANAL™ - Composite 2
∼1.11 g/mL
103°C