missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
HYDRANAL™ - Chloroform, Solvent for KF titration, Honeywell Fluka™
Solvent for Karl Fischer titration
$202.48 - $202.48
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
6000639
|
Honeywell Chemicals
378631L |
1 L |
Each for $202.48
Case of 6 Each for $1,214.88
|
|
|||||
Description
Advantages of HYDRANAL- Auxiliaries
- Broad product range of solubilizer for special sample application
- Broad product range of buffer for pH related sample application
- Drying agents for cell protection and drying of carrier gasses.
Chemical Identifiers
67-66-3 |
Specifications
1 L | |
Glass bottle | |
HYDRANAL™-Chloroform |
67-66-3,64-17-5 | |
UN1888 |
Safety and Handling
P260-P280-P301 + P312 + P330-P304 + P340 + P312-P305 + P351 + P338-P403 + P233
RTECSNumber : FS9100000