missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
HyClone™ RPMI 1640 Media
Designed to support cell culture growth and biomanufacturing processes.
Manufacturer: Ge Healthcare Bio-Science SH30027FS
Description
- Format: Liquid
Specifications
ISO 9001:2000 | |
RPMI 1640 media with L-glutamine | |
0.1 μm sterile filtered | |
L-glutamine |
Liquid | |
6 x 500 mL | |
2° to 8°C, away from light | |
2° to 8°C, away from light |