HyClone™ Fetal Bovine Serum (U.S.), Standard
Highly filtered and meets virus testing requirements.
Manufacturer:  Hyclone Laboratories Inc SH30088.03IH
Catalog No. SH3008803IH
Description
Description
Serum Collection:
  • From collection to final packaging
  • Every step is documented
  • Full traceability compliant with current Good Manufacturing Practices (cGMP)
Quick and Reliable Processing:
  • Extensive quality, performance, and analytical testing
  • Aseptically packaged in a Class 100 cleanroom
Quality Control Testing Standards:
  • Each lot of serum is tested to ensure quality and sterility
True Pool Processing:
  • Pooled after filtration and before final packaging
  • Consistent quality from the first bottle filled to the last
  • Increases consistency and minimizes variability with a lot of serum
Certifications
Manufactured under cGMP and ISO 9001 certified facilities.
Documents
FOR FURTHER MANUFACTURING OR RESEARCH USE. NOT FOR DIAGNOSTIC OR THERAPEUTIC USE.