missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Cytiva HyClone™ Fetal Bovine Serum (U.S.), Dialyzed
For applications that need serum without low molecular weight components.
$204.90 - $1326.69
Specifications
Country of Origin | United States |
---|---|
Specialty Type | Dialyzed |
Sterility | Sterile |
Sterilization Method | Sequential-filtered, 100 nm |
Grade | Specialty filtered |
Catalog Number | Mfr. No. | Quantity | Processing | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Processing | Price | Quantity | ||||
SH3007903
|
Cytiva
SH3007903 |
500 mL | None |
Each for $1,296.08
|
|
||||
SH3007901HI
|
Cytiva
SH3007901HI |
50 mL | Heat Inactivated |
Each for $204.90
|
|
||||
SH379IH2540
|
Cytiva
SH3007903IH2540 |
500 mL | Heat Inactivated, Gamma Irradiated (25 to 40 doses) |
Each for $1,326.69
|
|
||||
Description
Augment growth of cells with U.S.-sourced HyClone™ Dialyzed Fetal Bovine Serum (FBS), depleted of small molecules (less than 10,000 MW) and filtered through sequential 100nm (0.1μm) pore-size rated filters.
Specifications
United States | |
Sterile | |
Specialty filtered | |
≤ 10 mg/dL | |
USDA Inspected |
Dialyzed | |
Sequential-filtered, 100 nm | |
≤ 10 EU/mL | |
No |